ENA946831755 Enamine

5-carbamoyl-2-methylfuran-3-sulfonyl chloride

  • MDL number MFCD21196014
  • Cas Number : 1490847-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-277556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-277556
Availability details for
Quantity
  • Availability
  • Add to Cart