ENA946831759 Enamine

methyl 4-isocyanato-3-methylbenzoate

  • MDL number MFCD22557580
  • Cas Number : 864296-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-38
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-281529

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P312,P302+P352,P304+P340,P304+P341,P305+P351+P338,P312,P330,P332+P313,P337+P313,P342+P311,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-281529
pictograms GHS07,GHS08
hstatements H302,H312,H315,H319,H332,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart