ENA946831770 Enamine

ethyl cyclohexa-1,3-diene-1-carboxylate

  • MDL number MFCD00052374
  • Cas Number : 3725-40-4
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103289
Availability details for
Quantity
  • Availability
  • Add to Cart