ENA946831777 Enamine

4,4-dimethylhex-5-en-1-amine

  • MDL number MFCD19215267
  • Cas Number : 1007122-76-0
  • Molecular Weight : 127.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-300743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-300743
Availability details for
Quantity
  • Availability
  • Add to Cart