ENA946831798 Enamine

4-iodo-2-(1H-1,2,3,4-tetrazol-1-yl)benzoic acid

  • MDL number MFCD30730425
  • Cas Number : 2091778-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310335
Availability details for
Quantity
  • Availability
  • Add to Cart