ENA946831807 Enamine

3-({[1,2,4]triazolo[4,3-a]pyridin-3-yl}methyl)piperidine dihydrochloride

  • MDL number MFCD30730428
  • Cas Number : 2093718-99-9
  • Molecular Weight : 289.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-310943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-310943
Availability details for
Quantity
  • Availability
  • Add to Cart