ENA946831818 Enamine

2-(6-chloro-7-methyl-1-benzofuran-3-yl)acetic acid

  • MDL number MFCD30730430
  • Cas Number : 2094818-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-312327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-312327
Availability details for
Quantity
  • Availability
  • Add to Cart