ENA946831851 Enamine

methyl 1-[(5-cyanopyridin-2-yl)methyl]pyrrolidine-2-carboxylate

  • MDL number MFCD30730439

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315601
Availability details for
Quantity
  • Availability
  • Add to Cart