ENA946831856 Enamine

4-[(3-hydroxypyrrolidine-1-carbonyl)amino]oxane-4-carboxylic acid

  • MDL number MFCD25177956
  • Cas Number : 1556395-23-3
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-315990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315990
Availability details for
Quantity
  • Availability
  • Add to Cart