ENA946831863 Enamine

2-[(4-cyanophenyl)methoxy]acetic acid

  • MDL number MFCD26737617
  • Cas Number : 1361125-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-316242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-316242
Availability details for
Quantity
  • Availability
  • Add to Cart