ENA946831877 Enamine

1-cyclopropyl-2-methoxypropan-1-ol

  • MDL number MFCD21873678
  • Cas Number : 1539787-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317445
Availability details for
Quantity
  • Availability
  • Add to Cart