ENA946831881 Enamine

1-{5H,6H,7H,8H-pyrido[4,3-c]pyridazin-3-yl}pyrrolidine dihydrochloride

  • MDL number MFCD30730443
  • Cas Number : 2093582-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317552
Availability details for
Quantity
  • Availability
  • Add to Cart