ENA946831882 Enamine

oxane-2-carbonitrile

  • MDL number MFCD08669509
  • Cas Number : 5397-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317707
Availability details for
Quantity
  • Availability
  • Add to Cart