ENA946831883 Enamine

3,4-dihydrospiro[1-benzopyran-2,3'-piperidine] hydrochloride

  • MDL number MFCD30730444
  • Cas Number : 2094731-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317868
Availability details for
Quantity
  • Availability
  • Add to Cart