ENA946831884 Enamine

2,5-dicyclopropyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD30730445
  • Cas Number : 2094505-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-317946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-317946
Availability details for
Quantity
  • Availability
  • Add to Cart