ENA946831889 Enamine

2-(pyrazin-2-yl)propanenitrile

  • MDL number MFCD19671978
  • Cas Number : 1344351-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-318332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-318332
Availability details for
Quantity
  • Availability
  • Add to Cart