ENA946831893 Enamine

bis(N''-(3-methylbut-2-en-1-yl)guanidine); sulfuric acid

  • MDL number MFCD16293919
  • Cas Number : 14279-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-218
Purity 95
Salt Data 2H2SO4
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-320981

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-320981
pictograms GHS06
hstatements H301,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart