ENA946831901 Enamine

tert-butyl 7-amino-8,8-dimethyl-2-azabicyclo[4.2.0]octane-2-carboxylate

  • MDL number MFCD30730453
  • Cas Number : 2094316-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321320
Availability details for
Quantity
  • Availability
  • Add to Cart