ENA946831907 Enamine

2-chloro-6-(pentan-2-yloxy)pyridine

  • MDL number MFCD30730456
  • Cas Number : 1373770-41-2
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321824
Availability details for
Quantity
  • Availability
  • Add to Cart