ENA946831910 Enamine

2-(7-chloro-1H-indol-3-yl)acetonitrile

  • MDL number MFCD09924257
  • Cas Number : 221377-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321881
Availability details for
Quantity
  • Availability
  • Add to Cart