ENA946831914 Enamine

5-amino-1-(2-fluorophenyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD11054263
  • Cas Number : 630107-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322029
Availability details for
Quantity
  • Availability
  • Add to Cart