ENA946831919 Enamine

5-(chloromethyl)-4-methyl-2-phenyl-1,3-oxazole

  • MDL number MFCD11506350
  • Cas Number : 625106-10-7
  • Molecular Weight : 207.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322218
Availability details for
Quantity
  • Availability
  • Add to Cart