ENA946831922 Enamine

bis(1-methyl-1H-pyrazol-5-yl)methanamine

  • MDL number MFCD30730463
  • Cas Number : 1603462-95-8
  • Molecular Weight : 191.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322225
Availability details for
Quantity
  • Availability
  • Add to Cart