ENA946831923 Enamine

3-[(methylamino)methyl]pentan-3-ol

  • MDL number MFCD17218408
  • Cas Number : 227016-01-5
  • Molecular Weight : 131.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322230
Availability details for
Quantity
  • Availability
  • Add to Cart