ENA946831931 Enamine

1-[(3-chloro-2-methylpropyl)sulfanyl]-2-methylbenzene

  • MDL number MFCD19682425
  • Cas Number : 1341064-82-1
  • Molecular Weight : 214.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322249
Availability details for
Quantity
  • Availability
  • Add to Cart