ENA946831945 Enamine

3-fluoro-4-propoxyaniline

  • MDL number MFCD08687922
  • Cas Number : 946742-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322329
Availability details for
Quantity
  • Availability
  • Add to Cart