ENA946831947 Enamine

tert-butyl 1,2,3,4-tetrahydrophthalazine-2-carboxylate

  • MDL number MFCD24468352
  • Cas Number : 154972-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322334
Availability details for
Quantity
  • Availability
  • Add to Cart