ENA946832004 Enamine

5-methylpyrimidine-4,6-diamine

  • MDL number MFCD18819666
  • Cas Number : 162706-07-2
  • Molecular Weight : 124.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322529
Availability details for
Quantity
  • Availability
  • Add to Cart