ENA946832015 Enamine

2-phenyl-3,4-dihydro-2H-pyran-4-one

  • MDL number MFCD09032794
  • Cas Number : 40989-96-6
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-322587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-322587
Availability details for
Quantity
  • Availability
  • Add to Cart