ENA946832024 Enamine

4,7-dimethoxy-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD09736133
  • Cas Number : 64473-54-7
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135368
Availability details for
Quantity
  • Availability
  • Add to Cart