ENA946832045 Enamine

1-(benzyloxy)-3-fluoro-5-nitrobenzene

  • MDL number MFCD25019897
  • Cas Number : 1536920-33-8
  • Molecular Weight : 247.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332481
Availability details for
Quantity
  • Availability
  • Add to Cart