ENA946832050 Enamine

methyl 3-hydroxythiolane-3-carboxylate

  • MDL number MFCD16693865
  • Cas Number : 1247710-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333335
Availability details for
Quantity
  • Availability
  • Add to Cart