ENA946832051 Enamine

1,1,2-trichloroprop-1-ene

  • MDL number MFCD00040142
  • Cas Number : 21400-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139513

Safety Information

signalword Danger
pcodes P201,P202,P261,P264,P271,P273,P280,P281,P302+P352,P304+P340,P305+P351+P338,P308+P313,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-139513
pictograms GHS07,GHS08
hstatements H315,H319,H336,H341,H350,H412
Availability details for
Quantity
  • Availability
  • Add to Cart