ENA946832055 Enamine

3-(chloromethyl)-4,5-dihydro-1,2-oxazole

  • MDL number MFCD30730500
  • Cas Number : 1893432-12-6
  • Molecular Weight : 119.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-341838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-341838
Availability details for
Quantity
  • Availability
  • Add to Cart