ENA946832061 Enamine

(1S,3R,4S,5R)-3,4,5-trihydroxycyclohexane-1-carboxylic acid

  • MDL number MFCD30730502

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342174
Availability details for
Quantity
  • Availability
  • Add to Cart