ENA946832075 Enamine

1-[benzyl(methyl)amino]cyclohexane-1-carbonitrile

  • MDL number MFCD30617876
  • Cas Number : 41805-28-1
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343498
Availability details for
Quantity
  • Availability
  • Add to Cart