ENA946832078 Enamine

sodium 2-[(3S)-2,5-dioxo-2,3,4,5-tetrahydro-1H-1,4-benzodiazepin-3-yl]acetate

  • MDL number MFCD30730509
  • Cas Number : 2093997-56-7
  • Molecular Weight : 256.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343533
Availability details for
Quantity
  • Availability
  • Add to Cart