ENA946832085 Enamine

5-(methylamino)pyridine-2-carboxylic acid

  • MDL number MFCD24219430
  • Cas Number : 1499706-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343790
Availability details for
Quantity
  • Availability
  • Add to Cart