ENA946832090 Enamine

2,2-dimethyl-3,4-dihydro-2H-1-benzothiopyran-4-one

  • MDL number MFCD03066147
  • Cas Number : 28035-02-1
  • Molecular Weight : 192.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 63-65
Iupac Name 2,2-dimethyl-2,3-dihydro-4H-1-benzothiopyran-4-one
Inchi Code 1S/C11H12OS/c1-11(2)7-9(12)8-5-3-4-6-10(8)13-11/h3-6H,7H2,1-2H3
Inchi Key FJMCSSKIPWJNLP-UHFFFAOYSA-N

Description

Melting Point 63-65
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-344052

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-344052
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart