ENA946832096 Enamine

2-methyl-4-oxo-3,4-dihydroquinazoline-7-carboxylic acid

  • MDL number MFCD09815064
  • Cas Number : 773057-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-144826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-144826
Availability details for
Quantity
  • Availability
  • Add to Cart