ENA946832105 Enamine

2-bromo-1-isocyanato-4-methylbenzene

  • MDL number MFCD03701593
  • Cas Number : 71189-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-146861

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P285,P302+P352,P304+P340,P304+P341,P305+P351+P338,P312,P332+P313,P337+P313,P342+P311,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-146861
pictograms GHS08
hstatements H315,H319,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart