ENA946832107 Enamine

4,4-dimethylhexan-3-one

  • MDL number MFCD00048620
  • Cas Number : 19550-14-2
  • Molecular Weight : 128.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-345051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-345051
Availability details for
Quantity
  • Availability
  • Add to Cart