ENA946832136 Enamine

5-bromo-2,3-dihydrospiro[indene-1,3'-oxolane]-2',5'-dione

  • MDL number MFCD24149432
  • Cas Number : 1521827-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-349351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-349351
Availability details for
Quantity
  • Availability
  • Add to Cart