ENA946832139 Enamine

1-chloro-2-[(trifluoromethyl)sulfanyl]ethane

  • MDL number MFCD13194113
  • Cas Number : 819-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152317
Availability details for
Quantity
  • Availability
  • Add to Cart