ENA946832147 Enamine

1-methyl-4-{5H,6H,7H,8H-pyrido[4,3-c]pyridazin-3-yl}-1H-pyrazole trihydrochloride

  • MDL number MFCD30730536
  • Cas Number : 2093803-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-364707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-364707
Availability details for
Quantity
  • Availability
  • Add to Cart