ENA946832200 Enamine

1-(4-fluorophenyl)-3-(propan-2-yloxy)propan-2-one

  • MDL number MFCD12062096
  • Cas Number : 1158037-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-365795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365795
Availability details for
Quantity
  • Availability
  • Add to Cart