ENA946832202 Enamine

6-chloro-2,3-dihydro-1,2-benzothiazol-3-one

  • MDL number MFCD00865021
  • Cas Number : 70-10-0
  • Molecular Weight : 185.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-365814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-365814
Availability details for
Quantity
  • Availability
  • Add to Cart