ENA946832221 Enamine

1-(oxan-3-yl)propan-1-one

  • MDL number MFCD21193661
  • Cas Number : 1187468-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366052
Availability details for
Quantity
  • Availability
  • Add to Cart