ENA946832224 Enamine

3-(2-{[(tert-butoxy)carbonyl]amino}-1,3-thiazol-5-yl)propanoic acid

  • MDL number MFCD18822168
  • Cas Number : 920313-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366063
Availability details for
Quantity
  • Availability
  • Add to Cart