ENA946832231 Enamine

2-amino-5-cyclohexylthiophene-3-carbonitrile

  • MDL number MFCD23890925
  • Cas Number : 1505213-33-1
  • Molecular Weight : 206.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366110
Availability details for
Quantity
  • Availability
  • Add to Cart